tert-Butyl-N-(1,3-thiazol-2-yl)carbamate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: tert-Butyl thiazol-2-ylcarbamate

H58561-15 H58561-06
AAH58561-06EA 249.99 USD
AAH58561-06 AAH58561-15
tert-Butyl-N-(1,3-thiazol-2-yl)carbamate 97%
tert-Butyl-N-(1,3-thiazol-2-yl)carbamate
Formula: C₈H₁₂N₂O₂S
MW: 200.26 g/mol
Melting Pt: 180…185 °C
Storage Temperature: Ambient
MDL Number: MFCD09038327
CAS Number: 170961-15-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR