2-Amino-4-(2-pyridyl)thiazole 97%

Supplier: Thermo Scientific Chemicals

Warning

Synonyms: 4-(Pyridin-2-yl)thiazol-2-amine

H58554-04 H58554-09
AAH58554-09EA 1305.34 USD
AAH58554-09 AAH58554-04
2-Amino-4-(2-pyridyl)thiazole 97%
2-Amino-4-(2-pyridyl)thiazole
Formula: C₈H₇N₃S
MW: 177.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00460417
CAS Number: 30235-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR