Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.

4,5,6,7-Tetrahydro-1-benzothiophene-3-carboxylic acid ≥97%

Supplier: Alfa Aesar

H58548-06 H58548-03
AAH58548-06EA 524.23 USD InStock
AAH58548-06 AAH58548-03
4,5,6,7-Tetrahydro-1-benzothiophene-3-carboxylic acid ≥97%
4,5,6,7-Tetrahydro-1-benzothiophene-3-carboxylic acid


Formula: C₉H₁₀O₂S
MW: 182,24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00652575
CAS Number: 19156-54-8
Order Now

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty