2-Amino-5-chloro-6-methylpyridine 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 5-Chloro-6-methylpyridin-2-amine

H58501-06 H58501-03
AAH58501-06EA 516.99 USD
AAH58501-06 AAH58501-03
2-Amino-5-chloro-6-methylpyridine 97%
2-Amino-5-chloro-6-methylpyridine
Formula: C₆H₇ClN₂
MW: 142.59 g/mol
Melting Pt: 70…75 °C
Storage Temperature: Ambient
MDL Number: MFCD09453335
CAS Number: 36936-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR