2-(2-Thienyl)acetamidoxime 97%

Warning

H58358-09 H58358-04
AAH58358-09EA 809.19 USD
AAH58358-09 AAH58358-04
2-(2-Thienyl)acetamidoxime 97%
2-(2-Thienyl)acetamidoxime
Formula: C₆H₈N₂OS
Melting Pt: 68…74 °C
Storage Temperature: Ambient
MDL Number: MFCD01571357
CAS Number: 59174-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR