2-Amino-4-(5,6,7,8-tetrahydro-2-naphthyl)thiazole 97%

Warning

H58317-06
AAH58317-06EA 198.8 USD
AAH58317-06
2-Amino-4-(5,6,7,8-tetrahydro-2-naphthyl)thiazole 97%
2-Amino-4-(5,6,7,8-tetrahydro-2-naphthyl)thiazole
Formula: C₁₃H₁₄N₂S
MW: 230.33 g/mol
Melting Pt: 105…110 °C
Storage Temperature: Ambient
MDL Number: MFCD01804671
CAS Number: 87999-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR