4,6-Dichloro-5-methoxypyrimidine 97%

Danger

H58262-03 H58262-06 H58262-MD
AAH58262-06EA 261.23 USD
AAH58262-06 AAH58262-MD AAH58262-03
4,6-Dichloro-5-methoxypyrimidine 97%
4,6-Dichloro-5-methoxypyrimidine
Formula: C₅H₄Cl₂N₂O
MW: 179.01 g/mol
Melting Pt: 50…56 °C
Storage Temperature: Ambient
EINECS: 225-699-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR