N-(4-Methoxyphenyl)-3-nitrobenzenesulfonamide 97%

Supplier: Thermo Scientific Chemicals

Warning

H58131-ME H58131-MC
AAH58131-MEEA 289.71 USD
AAH58131-ME AAH58131-MC
N-(4-Methoxyphenyl)-3-nitrobenzenesulfonamide 97%
N-(4-Methoxyphenyl)-3-nitrobenzenesulphonamide
Formula: C₁₃H₁₂N₂O₅S
Storage Temperature: Ambient
MDL Number: MFCD00414861
CAS Number: 180631-81-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR