4-Fluoro-N,N-dimethylbenzamide 97%

Warning

Synonyms: N,N-Dimethyl 4-fluorobenzamide

H58080-MC H58080-ME
AAH58080-MCEA 36.33 USD
AAH58080-MC AAH58080-ME
4-Fluoro-N,N-dimethylbenzamide 97%
4-Fluoro-N,N-dimethylbenzamide
Formula: C₉H₁₀FNO
MW: 167.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01214152
CAS Number: 24167-56-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR