4-Bromo-N-(3-(trifluoromethyl)phenyl)benzamide 97%

Warning

H58046-ME H58046-MC
AAH58046-MCEA 88.17 USD
AAH58046-MC AAH58046-ME
4-Bromo-N-(3-(trifluoromethyl)phenyl)benzamide 97%
4-Bromo-N-(3-(trifluoromethyl)phenyl)benzamide
Formula: C₁₄H₉BrF₃NO
Storage Temperature: Ambient
MDL Number: MFCD00421306
CAS Number: 56661-32-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR