2-Methylisonicotinic acid ≥96%

Warning

Synonyms: 2-Methylpyridine-4-carboxylic acid

H57969-03 H57969-MD
AAH57969-MDEA 141.5 USD
AAH57969-MD AAH57969-03
2-Methylisonicotinic acid ≥96%
2-Methylisonicotinic acid
Formula: C₇H₇NO₂
MW: 137.14
Melting Pt: 295…300 °C
Storage Temperature: Ambient
MDL Number: MFCD00513435
CAS Number: 4021-11-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR