2-Chloro-N-methylnicotinamide 97%
|
Warning
|
H57963-03
H57963-MD
AAH57963-03EA
133.9
USD
AAH57963-03
AAH57963-MD
2-Chloro-N-methylnicotinamide 97%
2-Chloro-N-methylnicotinamide
Formula:
C₇H₇ClN₂O Storage Temperature: Ambient |
MDL Number:
MFCD08691934 CAS Number: 52943-20-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...