5-Chloro-3-methyl-1-phenyl-1H-pyrazole-4-carbaldehyde 97%

Supplier: Thermo Scientific Chemicals

Warning

Synonyms: 5-Chloro-3-methyl-1-phenyl-1H-pyrazole-4-carboxaldehyde

H57939-06 H57939-03
AAH57939-03EA 61.1 USD
AAH57939-03 AAH57939-06
5-Chloro-3-methyl-1-phenyl-1H-pyrazole-4-carbaldehyde 97%
5-Chloro-3-methyl-1-phenyl-1H-pyrazole-4-carbaldehyde
Formula: C₉H₁₁ClN₂O
MW: 220.65 g/mol
Melting Pt: 145…148 °C
Storage Temperature: Ambient
MDL Number: MFCD00100755
CAS Number: 947-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR