Oxetan-3-ol 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Hydroxyoxetane

H57928-03 H57928-06
AAH57928-03EA 35.19 USD
AAH57928-03 AAH57928-06
Oxetan-3-ol 95%
Oxetan-3-ol
Formula: C₃H₆O₂
MW: 74.08 g/mol
Boiling Pt: 153 °C
Density: 1.167
Flash Pt: 87 °C(189 °F)
Storage Temperature: Ambient
MDL Number: MFCD09056790
CAS Number: 7748-36-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR