(R)-Pyrrolidin-3-ylmethanol 95%

Supplier: Thermo Scientific Chemicals
Warning

H57871-MD H57871-03
AAH57871-03EA 384.99 USD
AAH57871-03 AAH57871-MD
(R)-Pyrrolidin-3-ylmethanol 95%
(R)-Pyrrolidin-3-ylmethanol
Formula: C₅H₁₁NO
MW: 101.15 g/mol
Boiling Pt: 173…177 °C
Density: 0.979
Storage Temperature: Ambient
MDL Number: MFCD09607969
CAS Number: 110013-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR