4-Methylpiperazin-2-one 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Methyl-3-oxopiperazine

H57633-03 H57633-MD
AAH57633-03EA 121.99 USD
AAH57633-03 AAH57633-MD
4-Methylpiperazin-2-one 95%
4-Methylpiperazin-2-one
Formula: C₅H₁₀N₂O
MW: 114.15 g/mol
Melting Pt: 84…90 °C
Storage Temperature: Ambient
MDL Number: MFCD11870124
CAS Number: 34770-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR