(S)-1-(tert-Butoxycarbonyl)-4-oxopiperidine-2-carboxylic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (S)-1-Boc-4-oxopipecolic acid(2S)-4-Oxopiperidine-2-carboxylic acid, N-BOC protected4-Oxo-L-pipecolinic acid, N-BOC protected(S)-(-)-1-Boc-4-oxopiperidine-2-carboxylic acid(2S)-1-(tert-Butoxycarbonyl)-4-oxopiperidine-2-carboxylic acid

H57617-03 H57617-MD
AAH57617-03EA 228.99 USD
AAH57617-03 AAH57617-MD
(S)-1-(tert-Butoxycarbonyl)-4-oxopiperidine-2-carboxylic acid 95%
(S)-1-(tert-Butoxycarbonyl)-4-oxopiperidine-2-carboxylic acid
Formula: C₁₁H₁₇NO₅
MW: 243.26 g/mol
Melting Pt: 121…126 °C
Storage Temperature: Ambient
MDL Number: MFCD01861764
CAS Number: 198646-60-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR