(S)-(+)-Nipecotic acid ≥96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: L-Nipecotic acid(S)-(+)-3-Piperidinecarboxylic acid(S)-Piperidine-3-carboxylic acid(S)-(+)-Piperidine-3-carboxylic acid

H57543-06 H57543-03 H57543-14
AAH57543-03EA 40.58 USD
AAH57543-03 AAH57543-14 AAH57543-06
(S)-(+)-Nipecotic acid ≥96%
(S)-(+)-Nipecotic acid
Formula: C₆H₁₁NO₂
MW: 129.16 g/mol
Melting Pt: 253…257 °C
Storage Temperature: Ambient
CAS Number: 59045-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR