4-Fluoro-N-(4-methylphenyl)benzamide 97%

Supplier: Thermo Scientific Chemicals

Warning

H57191-03 H57191-MD
AAH57191-03EA 318 USD
AAH57191-03 AAH57191-MD
4-Fluoro-N-(4-methylphenyl)benzamide 97%
4-Fluoro-N-(4-methylphenyl)benzamide
Formula: C₁₄H₁₂FNO
Storage Temperature: Ambient
MDL Number: MFCD00446355
CAS Number: 399-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR