2-Chloro-N,N-dimethylnicotinamide 97%

Supplier: Thermo Scientific Chemicals
Warning

H57162-MD H57162-03
AAH57162-03EA 127.99 USD
AAH57162-03 AAH57162-MD
2-Chloro-N,N-dimethylnicotinamide 97%
2-Chloro-N,N-dimethylnicotinamide
Formula: C₈H₉ClN₂O
Melting Pt: 63…65 °C
Storage Temperature: Ambient
MDL Number: MFCD08593346
CAS Number: 52943-21-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR