N-(4-Bromophenyl)-3-(trifluoromethyl)benzamide 97%

Supplier: Thermo Scientific Chemicals

Warning

H57015-MD H57015-03
AAH57015-MDEA 109.64 USD
AAH57015-MD AAH57015-03
N-(4-Bromophenyl)-3-(trifluoromethyl)benzamide 97%
N-(4-Bromophenyl)-3-(trifluoromethyl)benzamide
Formula: C₁₄H₉BrF₃NO
Storage Temperature: Ambient
MDL Number: MFCD03937564
CAS Number: 562080-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR