N-Benzyl-4-bromobenzenesulfonamide 97%

Warning

H56585-MD H56585-03
AAH56585-03EA 229.77 USD
AAH56585-03 AAH56585-MD
N-Benzyl-4-bromobenzenesulfonamide 97%
N-Benzyl-4-bromobenzenesulphonamide
Formula: C₁₃H₁₂BrNO₂S
MW: 326.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00450587
CAS Number: 3609-87-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR