4-(1H-Pyrazol-1-yl)benzoic acid tech. 90%, tech.

Supplier: Thermo Scientific Chemicals
Warning

H56405-06 H56405-03
AAH56405-06EA 420.99 USD
AAH56405-06 AAH56405-03
4-(1H-Pyrazol-1-yl)benzoic acid tech. 90%, tech.
4-(1H-Pyrazol-1-yl)benzoic acid
Formula: C₁₀H₈N₂O₂
MW: 188.18 g/mol
Melting Pt: 266 °C
Storage Temperature: Ambient
MDL Number: MFCD02682056
CAS Number: 16209-00-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR