5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na) 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: X-phosphate disodium saltSodium 5-bromo-4-chloro-1H-indol-3-yl phosphate

H56265-06 H56265-03
AAH56265-03EA 309.34 USD
AAH56265-03 AAH56265-06
5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na) 98+%
5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na)
Formula: C₈H₄BrClNNa₂O₄P
MW: 370.43 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00036757
CAS Number: 102185-33-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR