4-Fluoro-N-(4-fluorobenzyl)aniline 97%

Warning

H56208-03 H56208-MD
AAH56208-MDEA 145.41 USD
AAH56208-MD AAH56208-03
4-Fluoro-N-(4-fluorobenzyl)aniline 97%
4-Fluoro-N-(4-fluorobenzyl)aniline
Formula: C₁₃H₁₁F₂N
Storage Temperature: Ambient
MDL Number: MFCD03210766
CAS Number: 80143-73-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR