4-Hydroxyindanone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-HYDROXY-2,3-DIHYDRO-1H-INDEN-1-ONE4-hydroxyindan-1-one4-Hydroxy-1-indanone

H56184-06 H56184-03
AAH56184-03EA 150.98 USD
AAH56184-03 AAH56184-06
4-Hydroxyindanone 97%
4-Hydroxyindanone
Formula: C₉H₈O₂
MW: 148.161 g/mol
Melting Pt: 242…244 °C
Storage Temperature: Ambient
MDL Number: MFCD00143330
CAS Number: 40731-98-4
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR