1-(4-Methoxyphenylsulfonyl)pyrrolidine 97%

Warning

H56035-03 H56035-MD
AAH56035-03EA 283.05 USD
AAH56035-03 AAH56035-MD
1-(4-Methoxyphenylsulfonyl)pyrrolidine 97%
1-(4-Methoxyphenylsulphonyl)pyrrolidine
Formula: C₁₁H₁₅NO₃S
Storage Temperature: Ambient
MDL Number: MFCD00783727
CAS Number: 335215-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR