4-Fluoroindole 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Fluoro-1H-indole

H56011-06 H56011-03 H56011-MD
AAH56011-MDEA 83.79 USD
AAH56011-MD AAH56011-06 AAH56011-03
4-Fluoroindole 97%
4-Fluoroindole
Formula: C₈H₆FN
MW: 135.14 g/mol
Boiling Pt: 90 °C/0.4 mm
Melting Pt: 30…32 °C
Storage Temperature: Ambient
MDL Number: MFCD00055992
CAS Number: 387-43-9
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR