4-(Dimethylcarbamoyl)-2-fluorophenylboronic acid 97%

Warning

Synonyms: 4-Dimethylcarbamoyl-2-fluorobenzeneboronic acid

H55993-03 H55993-MD
AAH55993-MDEA 194.25 USD
AAH55993-MD AAH55993-03
4-(Dimethylcarbamoyl)-2-fluorophenylboronic acid 97%
4-(Dimethylcarbamoyl)-2-fluorophenylboronic acid
Formula: C₉H₁₁BFNO₃
Storage Temperature: Ambient
MDL Number: MFCD11855833
CAS Number: 874289-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR