2,5-Dibromo-3-dodecylthiophene 97%

H55958-06 H55958-MD H55958-03
AAH55958-03EA 105.67 USD
AAH55958-03 AAH55958-06 AAH55958-MD
2,5-Dibromo-3-dodecylthiophene 97%
2,5-Dibromo-3-dodecylthiophene
Formula: C₁₆H₂₆Br₂S
MW: 410.25 g/mol
Boiling Pt: 135 °C/2.5 mm
Density: 1.321
Flash Pt: >113 °C(235 °F)
Storage Temperature: Ambient
MDL Number: MFCD00274315
CAS Number: 148256-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR