5-Hydroxy-2-adamantanone 98%

Synonyms: 5-Hydroxyadamantan-2-one

H55950-03 H55950-MD H55950-06
AAH55950-03EA 85.99 USD
AAH55950-03 AAH55950-06 AAH55950-MD
5-Hydroxy-2-adamantanone 98%
5-Hydroxy-2-adamantanone
Formula: C₁₀H₁₄O₂
MW: 166.22 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00192211
CAS Number: 20098-14-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR