4-Carboxy-2-fluorophenylboronic acid 97%

Warning

Synonyms: 4-Carboxy-2-fluorobenzeneboronic acid

H55857-MD H55857-06 H55857-03
AAH55857-MDEA 66.52 USD
AAH55857-MD AAH55857-06 AAH55857-03
4-Carboxy-2-fluorophenylboronic acid 97%
4-Carboxy-2-fluorophenylboronic acid
Formula: C₇H₆BFO₄
MW: 183.93 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08458473
CAS Number: 851135-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR