3-Chloro-4-methylbenzonitrile 97%

Warning

H55833-06 H55833-03 H55833-14
AAH55833-03EA 26.59 USD
AAH55833-03 AAH55833-14 AAH55833-06
3-Chloro-4-methylbenzonitrile 97%
3-Chloro-4-methylbenzonitrile
Formula: C₈H₆ClN
MW: 151.60 g/mol
Melting Pt: 45…48 °C
Flash Pt: 84 °C(183 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001799
CAS Number: 21423-81-4
EINECS: 244-381-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR