2-Chloro-4-(methylcarbamoyl)phenylboronic acid 97%

Supplier: Thermo Scientific Chemicals

Warning

Synonyms: 2-Chloro-4-(methylcarbamoyl)benzeneboronic acid

H55778-03 H55778-MD
AAH55778-03EA 634 USD
AAH55778-03 AAH55778-MD
2-Chloro-4-(methylcarbamoyl)phenylboronic acid 97%
2-Chloro-4-(methylcarbamoyl)phenylboronic acid
Formula: C₈H₉BClNO₃
Storage Temperature: Ambient
MDL Number: MFCD22125174
CAS Number: 1451391-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR