5-Chloro-2-methylbenzothiazole 98%

H55743-14 H55743-06 H55743-03
AAH55743-06EA 54.75 USD
AAH55743-06 AAH55743-14 AAH55743-03
5-Chloro-2-methylbenzothiazole 98%
5-Chloro-2-methylbenzothiazole
Formula: C₈H₆ClNS
Melting Pt: 65…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00022881
CAS Number: 1006-99-1
EINECS: 213-746-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR