N-Methylbenzenesulfonamide 97%

Warning

H55666-03 H55666-MD
AAH55666-MDEA 42.57 USD
AAH55666-MD AAH55666-03
N-Methylbenzenesulfonamide 97%
N-Methylbenzenesulphonamide
Formula: C₇H₉NO₂S
MW: 171.21
Storage Temperature: Ambient
MDL Number: MFCD00025023
CAS Number: 5183-78-8
EINECS: 225-965-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR