trans-4-(4-Vinylcyclohexyl)benzonitrile 97%

Warning

Synonyms: 1-Cyano-4-(trans-4-vinylcyclohexyl)benzene4-(trans-4-Vinylcyclohexyl)benzonitrile

H55640-03 H55640-06
AAH55640-03EA 91.68 USD
AAH55640-03 AAH55640-06
trans-4-(4-Vinylcyclohexyl)benzonitrile 97%
trans-4-(4-Vinylcyclohexyl)benzonitrile
Formula: C₁₅H₁₇N
MW: 211.31 g/mol
Melting Pt: 57 °C
Storage Temperature: Ambient
MDL Number: MFCD11053490
CAS Number: 96184-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR