trans-4-(4-Pentylcyclohexyl)benzonitrile 97%

Warning

Synonyms: 4-(trans-4-n-Pentylcyclohexyl)benzonitrile

H55612-06 H55612-03
AAH55612-06EA 285.72 USD
AAH55612-06 AAH55612-03
trans-4-(4-Pentylcyclohexyl)benzonitrile 97%
trans-4-(4-Pentylcyclohexyl)benzonitrile
Formula: C₁₈H₂₅N
MW: 255.40 g/mol
Melting Pt: 30…55 °C
Flash Pt: 113 °C(235 °F)
Storage Temperature: Ambient
MDL Number: MFCD00075548
CAS Number: 61204-01-1
EINECS: 262-656-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR