Methyl-3-amino-2-methylbenzoate 96%

Warning

Synonyms: Methyl 3-amino-2-methylbenzoate

H55579-14 H55579-06 H55579-03
AAH55579-14EA 169.83 USD
AAH55579-14 AAH55579-03 AAH55579-06
Methyl-3-amino-2-methylbenzoate 96%
Methyl 3-amino-2-methylbenzoate
Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Boiling Pt: 300 °C
Density: 1.146
Flash Pt: 104 °C(219 °F)
Storage Temperature: Ambient
MDL Number: MFCD00211328
CAS Number: 18583-89-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR