2-Chloro-5-formylpyridine 96%

Supplier: Thermo Scientific Chemicals

Warning

Synonyms: 6-Chloronicotinaldehyde6-Chloropyridine-3-carboxaldehyde2-Chloropyridine-5-carboxaldehyde

H55550-03 H55550-06 H55550-MD
AAH55550-03EA 99.6 USD
AAH55550-03 AAH55550-06 AAH55550-MD
2-Chloro-5-formylpyridine 96%
2-Chloro-5-formylpyridine
Formula: C₆H₄ClNO
MW: 141.56 g/mol
Melting Pt: 77…81 °C
Storage Temperature: Ambient
MDL Number: MFCD03095223
CAS Number: 23100-12-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR