4-Bromo-o-anisaldehyde 97%

Warning

Synonyms: 4-Bromo-2-methoxybenzaldehyde

H55545-MD H55545-06 H55545-03
AAH55545-06EA 398.49 USD
AAH55545-06 AAH55545-MD AAH55545-03
4-Bromo-o-anisaldehyde 97%
4-Bromo-o-anisaldehyde
Formula: C₈H₇BrO₂
MW: 215.05 g/mol
Melting Pt: 67…71 °C
Storage Temperature: Ambient
MDL Number: MFCD08276823
CAS Number: 43192-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR