N-Benzyl-4-chlorobenzamide 97%

Warning

H55517-03 H55517-MD
AAH55517-03EA 462.87 USD
AAH55517-03 AAH55517-MD
N-Benzyl-4-chlorobenzamide 97%
N-Benzyl-4-chlorobenzamide
Formula: C₁₄H₁₂ClNO
Storage Temperature: Ambient
MDL Number: MFCD00018682
CAS Number: 7461-34-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR