Cyclopropanesulfonamide 97%
|
Warning
|
H55507-MD
H55507-06
H55507-03
AAH55507-03EA
202.37
USD
AAH55507-03
AAH55507-06
AAH55507-MD
Cyclopropanesulfonamide 97%
Cyclopropanesulphonamide
Formula:
C₃H₇NO₂S MW: 121.15 g/mol Melting Pt: 101…106 °C Storage Temperature: Ambient |
MDL Number:
MFCD08705286 CAS Number: 154350-29-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...