Methyl-2-aminothiophene-3-carboxylate 97%

Supplier: Thermo Scientific Chemicals

Synonyms: Methyl 2-aminothiophene-3-carboxylate

H55482-06 H55482-03
AAH55482-06EA 87.59 USD
AAH55482-06 AAH55482-03
Methyl-2-aminothiophene-3-carboxylate 97%
Methyl 2-aminothiophene-3-carboxylate
Formula: C₆H₇NO₂S
MW: 157.19 g/mol
Melting Pt: 76…81 °C
Storage Temperature: Ambient
MDL Number: MFCD00159547
CAS Number: 4651-81-4
EINECS: 225-084-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR