3-Octylthiophene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H55466-06
H55466-03
H55466-14
AAH55466-03EA
31.21
USD
AAH55466-03
AAH55466-14
AAH55466-06
3-Octylthiophene 97%
3-Octylthiophene
Formula:
C₁₂H₂₀S MW: 196.36 g/mol Boiling Pt: 106…107 °C/3 mm Density: 0.92 Flash Pt: 113 °C(235 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00085281 CAS Number: 65016-62-8 UN: 3334 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...