2,4-Difluorobenzenesulfonamide 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H55437-06
H55437-MD
H55437-03
AAH55437-06EA
472.99
USD
AAH55437-06
AAH55437-03
AAH55437-MD
2,4-Difluorobenzenesulfonamide 96%
2,4-Difluorobenzenesulphonamide
Formula:
C₆H₅F₂NO₂S MW: 193.17 g/mol Melting Pt: 154…158 °C Storage Temperature: Ambient |
MDL Number:
MFCD01320743 CAS Number: 13656-60-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...