8-(Tosylamino)quinoline 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 8-(p-Toluenesulphonylamino)quinoline

H55386-14 H55386-03 H55386-06
AAH55386-14EA 290.99 USD
AAH55386-14 AAH55386-03 AAH55386-06
8-(Tosylamino)quinoline 97%
8-(Tosylamino)quinoline
Formula: C₁₆H₁₄N₂O₂S
Melting Pt: 154…158 °C
Storage Temperature: Ambient
MDL Number: MFCD00168990
CAS Number: 10304-39-9

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR