trans-4-(4-n-Pentylcyclohexyl)phenyl-trans,trans-4'-n-propylbicyclohexyl-4-carboxylate 97%
Synonyms:
4-(trans-4-Pentylcyclohexyl)phenyl (trans,trans)-4'-Propyl-[1,1'-bi(cyclohexane)]-4-carboxylate
, (trans,trans)-4'-Propyl-[1,1'-bi(cyclohexane)]-4-carboxylic acid 4-(trans-4-Pentylcyclohexyl)phenyl ester
H55324-06
H55324-03
AAH55324-06EA
269.73
USD
AAH55324-06
AAH55324-03
trans-4-(4-n-Pentylcyclohexyl)phenyl-trans,trans-4'-n-propylbicyclohexyl-4-carboxylate 97%
trans-4-(4-n-Pentylcyclohexyl)phenyl-trans,trans-4'-n-propylbicyclohexyl-4-carboxylate
Formula:
C₃₃H₅₂O₂ MW: 480.777 g/mol Melting Pt: 213…223 °C Storage Temperature: Ambient |
MDL Number:
MFCD11053459 CAS Number: 131790-57-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...