1-(4-tert-Butylbenzyl)piperazine 97%

Warning

H55320-03 H55320-06 H55320-14
AAH55320-06EA 96.78 USD
AAH55320-06 AAH55320-03 AAH55320-14
1-(4-tert-Butylbenzyl)piperazine 97%
1-(4-tert-Butylbenzyl)piperazine
Formula: C₁₅H₂₄N₂
Melting Pt: 52…55 °C
Storage Temperature: Ambient
MDL Number: MFCD00082594
CAS Number: 956-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR