2-Amino-5-bromobenzothiazole 97%

Supplier: Thermo Scientific Chemicals
Warning

H55270-06 H55270-14 H55270-03
AAH55270-14EA 617.16 USD
AAH55270-14 AAH55270-03 AAH55270-06
2-Amino-5-bromobenzothiazole 97%
2-Amino-5-bromobenzothiazole
Formula: C₇H₅BrN₂S
MW: 229.1 g/mol
Melting Pt: 194…198 °C
Storage Temperature: Ambient
MDL Number: MFCD00577806
CAS Number: 20358-03-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR